EMAIL THIS PAGE TO A FRIEND
Biopolymers
Kinetics of chemical modification of arginine and lysine residues in calf thymus histone H1.
PMID 6793108
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M53701
o-Methylisourea bisulfate, 99%
C2H6N2O · H2SO4
455598
O-Methylisourea hemisulfate salt, 99%
C2H6N2O · 0.5 H2SO4