EMAIL THIS PAGE TO A FRIEND
The Indian journal of medical research
Bioavailability and pharmacokinetics of chloramphenicol palmitate in malnourished children.
PMID 6796509
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46109
Chloramphenicol palmitate, VETRANAL, analytical standard
C27H42Cl2N2O6