EMAIL THIS PAGE TO A FRIEND
Nihon Ketsueki Gakkai zasshi : journal of Japan Haematological Society
The analysis of platelet reaction, utilizing various inhibitors of platelet aggregation.
PMID 6800180