EMAIL THIS PAGE TO A FRIEND
Comptes rendus des seances de l'Academie des sciences. Serie III, Sciences de la vie
[Effect of thyroid deficiency on cutaneous nerve development in chickens].
PMID 6804024
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
115169
Tetramethylthiourea, 98%
C5H12N2S