EMAIL THIS PAGE TO A FRIEND
Acta histochemica. Supplementband
[A combined carbohydrate immunocytochemical technic for localization of glycogen in the pancreatic islands of steroid diabetic rats].
PMID 6806863
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185191
Lead(IV) acetate, reagent grade, 95%
C8H12O8Pb