EMAIL THIS PAGE TO A FRIEND
The International journal of biochemistry
Beta-Glucosidase: its role in cellulase synthesis and hydrolysis of cellulose.
PMID 6809504
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49290
β-Glucosidase from almonds, lyophilized, powder, ≥4 U/mg