EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
Benzal chloride.
PMID 6813242
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251046
α,α-Dichlorotoluene, 95%
C7H6Cl2