EMAIL THIS PAGE TO A FRIEND
Neurotoxicology
Dithiobiuret-induced muscle weakness in rats: evidence for a prejunctional effect.
PMID 6813777
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3190
Dithiobiuret, 97%, solid