EMAIL THIS PAGE TO A FRIEND
Diabetes
Plasma prostaglandin levels in rats with diabetes mellitus and diabetic ketoacidosis.
PMID 6816644
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
644927
3-Methylpyrazole-5-carboxylic acid, 97%
C5H6N2O2