EMAIL THIS PAGE TO A FRIEND
Hemoglobin
Detection of Hb-Papio B, a silent mutation of the baboon beta chain, by high performance liquid chromatography. Improved procedures for the separation of globin chains by HPLC.
PMID 6816761
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74398
Nonylamine, ≥99.5% (GC)
C9H21N
N31001
Nonylamine, 98%
C9H21N