EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
De novo formation of the niaD gene directed protomer in the NADPH-nitrate reductase of Aspergillus nidulans.
PMID 6817753