EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A calorimetric investigation of the binding of indole and phenylethane boronic acid to chymotrypsin.
PMID 6822550
Abstract
The heat of formation of the chymotrypsin-phenylethane boronic acid complex has been observed calorimetrically from pH 4 to 8 at 25 degrees C and is found to be pH-dependent, changing from near -6 kcal/mol at pH 4 to -13 kcal/mol at pH 8. The heat of formation of the chymotrypsin-indole complex is a nearly constant -6 kcal/mol over most of the same pH range. alpha-Chymotrypsin has been purified by pH gradient elution from an immobilized lima bean inhibitor column. Solutions of the enzyme up to 400 microM, prepared in this manner, have a zero heat of dilution from pH 5 to 8 in 0.1 M KCl, with or without added 0.05 M Tris, N-(tris[hydroxy-methyl]methyl-2-amino) ethanesulfonic acid, 4-morpholineethanesulfonic acid, or acetate buffers. Binding of phenylethane boronic acid causes a pH-dependent decrease in proton binding to chymotrypsin; the decrease in proton binding evoked by formation of the indole complex is much less, with a much smaller pH dependence. The calorimetric and proton-binding results are applied to a model for boronic acid binding (Hanai, K. (1976) J. Biochem. (Tokyo) 79, 107-116). We conclude that the thermodynamics of formation of the trigonal boronic acid complex are quite similar to those for the formation of the noncovalent complex formed by indole and related ligands. The trigonal-tetrahedral tautomerism in the boronic acid-chymotrypsin complex is characterized by thermodynamic changes similar to those accompanying the binding of virtual substrates to chymotrypsin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
588423
Phenethylboronic acid
C8H11BO2