EMAIL THIS PAGE TO A FRIEND
Cancer research
LNCaP model of human prostatic carcinoma.
PMID 6831420
Abstract
The LNCaP cell line was established from a metastatic lesion of human prostatic adenocarcinoma. The LNCaP cells grow readily in vitro (up to 8 x 10(5) cells/sq cm; doubling time, 60 hr), form clones in semisolid media, are highly resistant to human fibroblast interferon, and show an aneuploid (modal number, 76 to 91) human male karyotype with several marker chromosomes. The malignant properties of LNCaP cells are maintained. Athymic nude mice develop tumors at the injection site (volume-doubling time, 86 hr). Functional differentiation is preserved; both cultures and tumor produce acid phosphatase. High-affinity specific androgen receptors are present in the cytosol and nuclear fractions of cells in culture and in tumors. Estrogen receptors are demonstrable in the cytosol. The model is hormonally responsive. In vitro, 5 alpha-dihydrotestosterone modulates cell growth and stimulates acid phosphatase production. In vivo, the frequency of tumor development and the mean time of tumor appearance are significantly different for either sex. Male mice develop tumors earlier and at a greater frequency than do females. Hormonal manipulations show that, regardless of sex, the frequency of tumor development correlates with serum androgen levels. The rate of the tumor growth, however, is independent of the gender of hormonal status of the host.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89110211
LNCap Clone FGC Cell Line human, from human prostate(carcinoma)