EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and antiallergic activity of some acidic derivatives of 4H-pyrimido[2,1-b]benzazol-4-ones.
PMID 6834395
Abstract
Reactions of 2-aminobenzothiazole, 2-aminobenzoxazole, and 2-amino-1-methylbenzimidazole with dimethyl aminofumarate (DMAF) or diethyl ethoxymethylenemalonate (DEEM) led to 2- or 3-carboxy-4H-pyrimido[2,1-b]-benzazol-4-ones, respectively. Subsequent derivatization of these carboxylic acids gave the corresponding tetrazolylcarboxamides and tetrazoles. These acidic compounds were tested in the rat passive cutaneous anaphylaxis (PCA) assay as potential antiallergic agents. Many of the compounds displayed activity comparable to that shown by disodium cromoglycate (DSCG) when tested by the intraperitoneal route, and some, unlike DSCG, also showed activity when tested orally.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
412546
2-Amino-1-methylbenzimidazole, 95%
C8H9N3