EMAIL THIS PAGE TO A FRIEND
Drug and therapeutics bulletin
Co-trifamole (Co-Fram).
PMID 6839969
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46866
Sulfamoxole, VETRANAL®, analytical standard
C11H13N3O3S