EMAIL THIS PAGE TO A FRIEND
Meditsinskaia parazitologiia i parazitarnye bolezni
[Comparative effectiveness of 2 types of baits for synanthropic flies in different regions].
PMID 6843498
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45450
Dimetilan, PESTANAL®, analytical standard
C10H16N4O3