EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
(-)-10,11-Methylenedioxy-N-propylnoraporphine, an orally effective dopamine agonist and duodenal antiulcerogen in the rat.
PMID 6852114
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M121
(−)-MDO-NPA hydrochloride
C20H21NO2 · xHCl