EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Determination of total protein.
PMID 6855607
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47641 Folin & Ciocalteu’s phenol reagent, 2 M (with respect to acid)