EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Sensitivity and specificity of eicosanoid radioimmunoassays: new strategy.
PMID 6860583
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3631
Arachidic acid, ≥99%
C20H40O2
10930
Arachidic acid, synthetic, ≥99.0% (GC)
C20H40O2