EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Kinetics and mechanism of hydroxy compound cinnamoylation in acetonitrile catalyzed by N-methylimidazole and 4-dimethylaminopyridine.
PMID 6864471
Abstract
The kinetics of reaction of the acylating agents trans-cinnamic anhydride and trans-cinnamoyl chloride with the hydroxy compounds n-propyl alcohol and water in the presence of N-methylimidazole and 4-dimethylaminopyridine were studied spectrophotometrically in acetonitrile solution at 25 degrees. The acid chloride reacted via the intermediate formation of the N-acyl catalyst, which underwent general base-catalyzed reaction with the hydroxy compound. The anhydride did not form the N-acyl intermediate, but instead underwent direct general base catalysis. In the presence of water, all systems formed the N-acyl intermediate. The mechanistic route followed by the system was determined by the nucleophilicity of the catalyst, the ability of the leaving group, and the polarity of the solvent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C81101
Cinnamoyl chloride, 98%
C9H7ClO