EMAIL THIS PAGE TO A FRIEND
Journal of animal science
Effects of lasalocid or monensin on legume or grain (feedlot) bloat.
PMID 6874619
Abstract
Doses of .66 to .99 mg monensin/kg body weight reduced legume bloat in cattle about 66% when compared with pretreatment bloat scores. Similar doses of lasalocid reduced legume bloat about 26%. A dose of 44 mg poloxalene/kg body weight (recommended dose for field use) reduced legume bloat 100%. Monensin or lasalocid combined with 25 or 50% of the recommended dose of poloxalene reduced bloat under that of the antibiotics alone, but did not achieve 100% reduction. The antibiotic thiopeptin provided no preventive effect on legume bloat. Lasalocid, monensin or an experimental polyether antibiotic (X-14,547 A) at a dose of 1.32 mg/kg body weight when tested on cattle bloated on high grain diets reduced bloat by 92, 64 and 25%, respectively. Lasalocid at .66 mg/kg effectively prevented bloat from developing when given to animals before the feeding of high grain diets; however, a 1.32-mg dose was required to control bloat in cattle that were already bloating before they were given lasalocid. A dose of 1.32 mg salinomycin was ineffective in controlling grain bloat.