EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
The fractional inhibitory concentration (FIC) index as a measure of synergy.
PMID 6874629
Abstract
The value of the FIC index as a predictor of synergy has been investigated using the antibacterial agents alafosfalin and cephalexin combined together with themselves in fully blind experiments. Under the conditions used, even weak interaction (FIC index 0.5-0.99) proved to be statistically highly significant. The use of such fully controlled blind studies would greatly enhance the credibility of many of the claims of synergy published in the literature. The representation of results as average isobolograms is only of value with combinations which show moderate to strong interaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05260
L-Alanyl-L-1-aminoethylphosphonic acid, ≥98.0% (T)
C5H13N2O4P