EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
A new class of nonhormonal pregnancy-terminating agents. Synthesis and contragestational activity of 3,5-diaryl-s-triazoles.
PMID 6876087
Abstract
A series of 3,5-diaryl-s-triazoles were synthesized and evaluated as postimplantation contragestational agents. The introduction of various substituents (e.g., an o-alkyl chain on one phenyl and a m-alkoxy group on the other) was found to increase the potency. Several compounds with very high pregnancy-terminating activity in both hamsters and rats were obtained. One of these, 3-(2-ethylphenyl)-5-(3-methoxyphenyl)-s-triazole, DL 111 (36), was selected for detailed evaluation in various animal species. A synthetic scheme for the preparation of these compounds and preliminary structure-activity relationships are presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
525049
4-Methoxy-2-methylbenzoic acid, 97%
C9H10O3