EMAIL THIS PAGE TO A FRIEND
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences
[Studies on analysis of pesticide residues in foods (XXXVIII). Surveillance of chlorinated phenoxy herbicide residues in imported agricultural crops in 1980].
PMID 6892148