EMAIL THIS PAGE TO A FRIEND
Meditsinskaia parazitologiia i parazitarnye bolezni
[The effect of fenbendazole and parbendazole on the course of experimental trichinelliasis in white mice].
PMID 6894480
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32438
Parbendazole, VETRANAL, analytical standard
C13H17N3O2