EMAIL THIS PAGE TO A FRIEND
Biochemistry
Lanthanide probes in biological systems: the calcium binding site of pancreatic elastase as studied by terbium luminescence.
PMID 6899954
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5510
N-Acetyl-Ala-Ala-Ala
C11H19N3O5