EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
The effect of low-molecular-weight inhibitors on the growth in vitro of a human pancreatic adenocarcinoma cell line.
PMID 6908576
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0881
Elastatinal, microbial
C21H36N8O7