EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Acetaldehyde, methanol, and ethanol analysis by headspace gas chromatography.
PMID 6927051
Abstract
This report describes a headspace procedure suitable for the gas chromatographic assay of acetaldehyde, methanol, ethanol and various other volatile substances in biological samples. Carbopack B coated with 5% Carbowax 20-M is compared to Carbopack C coated with 0.2% Carbowax 1500. Assay range, linearity and precision are reported for the Carbopack B material.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20273 Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
10257 Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
10258 Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
10436 Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
10437-U Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
10439-U Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
10461-U Carbopack Adsorbent, matrix Carbopack Y, 40-60 mesh, bottle of 10 g
10462 Carbopack Adsorbent, matrix Carbopack Y, 60-80 mesh, bottle of 10 g
10464-U Carbopack Adsorbent, matrix Carbopack Y, 120-400 mesh, bottle of 50 g
11051-U Carbopack Adsorbent, matrix Carbopack Z, 60-80 mesh, bottle of 10 g
29535-U Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
30136-U Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
30135-U
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
29754-U Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29537-U Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
29756-U Carbopack X, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
11809 Coated GC Packing, phase 1% SP-1510, matrix 60/80 Carbopack B support, bottle of 15 g
28686-U
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)