EMAIL THIS PAGE TO A FRIEND
Caries research
Metabolism of glucosylsucrose and maltosylsucrose by Streptococcus mutans.
PMID 6931633
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1895
Erlose
C18H32O16