EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Agonists of Torpedo nicotinic receptors: essential role of a positive charge.
PMID 6938159
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2762
(Dansylaminoethyl)­tri­methyl­ammonium perchlorate
C17H26N3O2S · ClO4