EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Micropinocytic ingestion of glycosylated albumin by isolated microvessels: possible role in pathogenesis of diabetic microangiopathy.
PMID 6941299
Abstract
Microvessels isolated from rat epididymal fat exhibit differential vesicular ingestion rates for unmodified and non-enzymatically glycosylated rat albumin. While unmodified rat albumin is excluded from ingestion by endothelial micropinocytic vesicles, glycosylated albumin is avidly taken up by endocytosis. Interaction of albumin and glycosylated albumin with endothelium was studied with a double-label fluorescence assay of micropinocytosis. When glycosylated albumin was present at a concentration of 6% with respect to total albumin (the level found in "non diabetic" serum), only glycosylated albumin was ingested. At higher concentrations of glycosylated albumin (those found in diabetic serum), both albumin and glycosylated albumin are ingested. Glycosylation of endothelial membrane components results in stimulated ingestion of glycosylated albumin, persistent exclusion of unmodified albumin, and unaltered micropinocytic ingestion of native ferritin. These results indicate that nonenzymatic glycosylation of serum albumin may result in rapid vesicle-mediated extravasation of albumin. Chronic microvascular leakage of glycosylated albumin could contribute to the pathogenesis of diabetic microangiopathy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R1755
Rhodamine B isothiocyanate, BioReagent, mixed isomers, suitable for protein labeling
C29H30ClN3O3S