EMAIL THIS PAGE TO A FRIEND
Shika gakuho. Dental science reports
[Study on the effect of tellurium for experimental dental caries development in rat. On the effects of sodium tellurite and glutathione combination for nutrition and experimental dental caries development (author's transl)].
PMID 6946565
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
400688
Sodium tellurite, −100 mesh, 99%
Na2O3Te