EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Partial purification and characterization of two isoenzymes involved in the sulfurylation of catecholamines.
PMID 6956338