EMAIL THIS PAGE TO A FRIEND
Acta chirurgica Scandinavica. Supplementum
Methodological considerations on antithrombin determination.
PMID 6962626
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7632
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, protease substrate
C33H40N8O6 · HCl