EMAIL THIS PAGE TO A FRIEND
Nihon Shishubyo Gakkai kaishi
[Clinical effect of dentifrice containing dl-alpha-Tocopheryl nicotinate].
PMID 6963312
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5134
(±)-α-Tocopherol nicotinate
C35H53NO3