EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The effect of nitrous oxide-induced inactivation of vitamin B12 on the activity of formyl-methenyl-methylenetetrahydrofolate synthetase, methylene-tetrahydrofolate reductase and formiminotetrahydrofolate transferase.
PMID 6971097