EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Completion of the amino acid sequences of the A and B chains of subcomponent C1q of the first component of human complement.
PMID 6981411
Abstract
The sequences of amino acid residues 109--224 of the A chain, and residues 109--22 of the B chain, of human subcomponent C1q are given. These results, along with previously published sequence data on the N-terminal, collagen-like, regions of the A and B chains [Reid (1979) Biochem. J. 179, 367--371] yield the complete amino acid sequences of the A and B chains of subcomponent C1q. The asparagine residue at position A-124 has been identified as the major site of asparagine-linked carbohydrate in subcomponent C1q. When the sequences of the C-terminal, 135-residue-long, 'globular' regions of A and B chains are compared they show 40% homology. The degree of homology over certain stretches of 15--20 residues, within the C-terminal regions, rises up to values of 73%, indicating the presence of strongly conserved structures. Structure prediction studies indicate that both the A and B chain C-terminal regions may adopt a predominantly beta-type structure with apparently little alpha-helical structure.