EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Electrophoresis and electrofocusing on polyacrylamide gel in the study of native macromolecules.
PMID 6988692
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
156868
(+)-N,N′-Diallyltartramide, ≥99%
C10H16N2O4