EMAIL THIS PAGE TO A FRIEND
Annual review of biochemistry
Blood coagulation.
PMID 6996572
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9302
Factor X Activated (Xa) from bovine plasma, aqueous glycerol solution