EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Identification of an intermediate in the de novo formation of nicotinamide adenine dinucleotide in Escherichia coli.
PMID 6998482