EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Fennel and anise as estrogenic agents.
PMID 6999244
Abstract
Fennel, Foeniculum vulgare, and anise, Pimpinella anisum, are plants which have been used as estrogenic agents for millennia. Specifically, they have been reputed to increase milk secretion, promote menstruation, facilitate birth, alleviate the symptoms of the male climacteric, and increase libido. In the 1930s, some interest was shown in these plants in the development of synthetic estrogens. The main constituent of the essential oils of fennel and anise, anethole, has been considered to be the active estrogenic agent. However, further research suggests that the actual pharmacologically active agents are polymers of anethole, such as dianethole and photoanethole.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W248207 Fennel oil, FCC, FG