EMAIL THIS PAGE TO A FRIEND
Farmakologiia i toksikologiia
[Tranquilizing activity of adamantane derivatives].
PMID 7002591
Abstract
It was established in experiments on mice and rats carried out according to conventional procedures that 13 adamantane derivatives have a greater tranquilizing activity than amantadine, 3-bromadamantoyl-1 diazomethane exerting a stronger effect than trioxazin. Two adamantane derivatives were found to have an anticonvulsant activity.