EMAIL THIS PAGE TO A FRIEND
Mikroskopie
[Photographic models in the study and valuation of the silver reactions in histology. II. Optical density variation in enlarging exposure scale under the action of the ammonium sulphide on silver (author's transl)].
PMID 7003437