EMAIL THIS PAGE TO A FRIEND
Essays in neurochemistry and neuropharmacology
Stereospecific transport carrier systems for L-tyrosine and L-tryptophan in relation to monoamine synthesis.
PMID 7014213
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1705006
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
C9H11NO3