EMAIL THIS PAGE TO A FRIEND
Cutis
Allergic contact dermatitis in a violinist. The role of abietic acid--a sensitizer in rosin (colophony)--as the causative agent.
PMID 7016461
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00010
Abietic acid, technical, ~75% (GC)
C20H30O2