EMAIL THIS PAGE TO A FRIEND
Gigiena i Sanitariia
[Hygienic importance of the transformation of chemical substances with the formation of chloroform in the process of drinking water decontamination].
PMID 7024047