EMAIL THIS PAGE TO A FRIEND
IARC scientific publications
A survey of techniques for estimating and monitoring aromatic amines of major industrial importance.
PMID 7035352
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46106
4,4′-Methylene-bis(2-methylaniline), analytical standard
C15H18N2