EMAIL THIS PAGE TO A FRIEND
The Japanese journal of antibiotics
[Review [new antibiotics series V]: cefaclor (author's transl)].
PMID 7047791
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6895
Cefaclor
C15H14ClN3O4S