EMAIL THIS PAGE TO A FRIEND
American Industrial Hygiene Association journal
Hexadiene.
PMID 7055078
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129186
1,4-Hexadiene, mixture of cis and trans, 99%
C6H10