EMAIL THIS PAGE TO A FRIEND
Biopolymers
Fluorescent-labeled crosslinks in collagen: pyrenebutyrylhydrazine.
PMID 7055636
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82669
1-Pyrenebutyric hydrazide, for fluorescence, ≥97.0% (T)
C20H18N2O